2-Fluoro-6-iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15138-25G 15138-5G
76895-672EA 233.4 USD
76895-672 76895-674
2-Fluoro-6-iodophenylboronic acid ≥97%
2-Fluoro-6-iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BFIO₂
Storage Temperature: Ambient
MDL Number: MFCD06798080
CAS Number: 870777-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR