2,6-Difluoro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,6-Difluoro-3-formylphenyl)boronic acid

15122-1G 15122-25G 15122-5G
76895-660EA 88.06 USD
76895-660 76895-662 76895-664
2,6-Difluoro-3-formylphenylboronic acid ≥97%
2,6-Difluoro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₃
MW: 185.9200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369756
CAS Number: 849062-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR