3-(1,3,2-Dioxaborinan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

149978-25G 149978-5G
76895-634EA 428.61 USD
76895-634 76895-636
3-(1,3,2-Dioxaborinan-2-yl)phenol ≥97%
3-(1,3,2-Dioxaborinan-2-yl)phenol
Formula: C₉H₁₁BO₃
MW: 177.9928 g/mol
Storage Temperature: Refrigerator
CAS Number: 2070921-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR