2-Chloro-1-ethyl-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

148275-1G 148275-25G 148275-500MG 148275-5G
76895-604EA 233.4 USD
76895-604 76895-606 76895-608 76895-610
2-Chloro-1-ethyl-3-methylbenzene ≥97%
2-Chloro-1-ethyl-3-methylbenzene
Formula: C₉H₁₁Cl
Storage Temperature: Ambient
CAS Number: 344612-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR