2-Formyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

14766-1G 14766-25G 14766-500MG 14766-5G
76895-594EA 201.58 USD
76895-594 76895-596 76895-598 76895-600
2-Formyl-4-methoxybenzoic acid ≥97%
2-Formyl-4-methoxybenzoic acid
Formula: C₉H₈O₄
MW: 180.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18453659
CAS Number: 60373-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR