≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dibromo-4-methoxytoluene, 1,3-Dibromo-2-methoxy-5-methylbenzene, 2,6-Dibromo-4-Methylanisole

14566-25G 14566-5G
76895-572EA 346.92 USD
76895-572 76895-574
≥95%
2,6-Dibromo-4-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.96
Storage Temperature: Ambient
MDL Number: MFCD00196821
CAS Number: 51699-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR