2-Bromo-5-iodothiophene ≥97%

Supplier: AOBChem USA

Warning

14549-1G 14549-25G 14549-5G
76895-562EA 76.39 USD
76895-562 76895-564 76895-566
2-Bromo-5-iodothiophene ≥97%
2-Bromo-5-iodothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrIS
Storage Temperature: Refrigerator
MDL Number: MFCD08695058
CAS Number: 29504-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR