2-Fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14535-1G 14535-25G 14535-5G
76895-542EA 66.84 USD
76895-542 76895-544 76895-546
2-Fluoro-3-hydroxybenzaldehyde ≥97%
2-Fluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD11110191
CAS Number: 103438-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR