(2-Formyl-4-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Formyl-4-methylphenylboronic acid

14491-1G 14491-25G 14491-500MG 14491-5G
76895-510EA 201.58 USD
76895-510 76895-512 76895-514 76895-516
(2-Formyl-4-methylphenyl)boronic acid ≥97% research grade
(2-Formyl-4-methylphenyl)boronic acid
Formula: C₈H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD13181634
CAS Number: 444188-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR