2-Hydroxy-5-(trifluoromethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

14475-25G 14475-5G
76895-490EA 595.17 USD
76895-490 76895-492
2-Hydroxy-5-(trifluoromethoxy)phenylboronic acid ≥95%
2-Hydroxy-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₄
MW: 221.926 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397559
CAS Number: 1354819-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR