2-Fluoro-3-hydroxybenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

14453-25G 14453-5G
76895-472EA 321.46 USD
76895-472 76895-474
2-Fluoro-3-hydroxybenzeneboronic acid ≥97%
2-Fluoro-3-hydroxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
Storage Temperature: Ambient
MDL Number: MFCD06797222
CAS Number: 855230-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR