(2-Fluoro-6-isopropoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluro-6-isopropoxyphenylboronic acid

14416-25G 14416-5G
76895-462EA 275.84 USD
76895-462 76895-464
(2-Fluoro-6-isopropoxyphenyl)boronic acid ≥97%
(2-Fluoro-6-isopropoxyphenyl)boronic acid
Formula: C₉H₁₂BFO₃
MW: 197.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798078
CAS Number: 870777-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR