2-Chloro-(5-ethoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-(ethoxycarbonyl)phenylboronic acid, (2-Chloro-5-(ethoxycarbonyl)phenyl)boronic acid

14379-1G 14379-25G 14379-5G
76895-454EA 70.02 USD
76895-454 76895-456 76895-458
2-Chloro-(5-ethoxycarbonyl)phenylboronic acid ≥97%
2-Chloro-(5-ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BClO₄
Storage Temperature: Ambient
MDL Number: MFCD08689529
CAS Number: 913835-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR