5-Carboxy-2-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-carboxyphenylboronic acid

14377-25G 14377-5G
76895-444EA 219.61 USD
76895-444 76895-446
5-Carboxy-2-chlorophenylboronic acid ≥97%
5-Carboxy-2-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₄
MW: 200.3800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689491
CAS Number: 913835-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR