2,6-Dichloropyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dichloroisonicotinaldehyde

14341-1G 14341-25G 14341-5G
76895-434EA 88.06 USD
76895-434 76895-436 76895-438
2,6-Dichloropyridine-4-carboxaldehyde ≥97%
2,6-Dichloropyridine-4-carboxaldehyde
Formula: C₆H₃Cl₂NO
MW: 176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00124816
CAS Number: 113293-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR