2-Chloro-3-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

14220-25G 14220-5G
76895-386EA 339.49 USD
76895-386 76895-388
2-Chloro-3-fluoro-4-iodopyridine ≥97%
2-Chloro-3-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClFIN
MW: 257.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972389
CAS Number: 148639-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR