2-Fluoro-4-iodo-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-iodo-5-picoline

14210-25G 14210-5G
76895-374EA 204.76 USD
76895-374 76895-376
2-Fluoro-4-iodo-5-methylpyridine ≥97%
2-Fluoro-4-iodo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095297
CAS Number: 153034-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR