6-Fluoro-5-methylnicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-methyl-pyridine-5-carboxylic acid

14192-25G 14192-5G
76895-346EA 418 USD
76895-346 76895-348
6-Fluoro-5-methylnicotinic acid ≥97% research grade
6-Fluoro-5-methylnicotinic acid
Formula: C₇H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD07781235
CAS Number: 885267-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR