6-Bromo-5-methylnicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-formyl-3-methylpyridine

14146-1G 14146-25G 14146-500MG 14146-5G
76895-316EA 119.89 USD
76895-316 76895-318 76895-320 76895-322
6-Bromo-5-methylnicotinaldehyde ≥95%
6-Bromo-5-methylnicotinaldehyde
Formula: C₇H₆BrNO
MW: 200.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08277302
CAS Number: 885167-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR