2-Ethoxypyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxypyridine-3-carbaldehyde, 2-Ethoxynicotinaldehyde

14143-1G 14143-25G 14143-5G 14143-500MG
76895-302EA 114.58 USD
76895-302 76895-304 76895-306 77648-634
2-Ethoxypyridine-3-carboxaldehyde ≥97%
2-Ethoxypyridine-3-carboxaldehyde
Formula: C₈H₉NO₂
MW: 151.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06804559
CAS Number: 885278-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR