2-Chloro-5-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-methylpyridine-3-carbaldehyde

14130-1G 14130-25G 14130-500MG 14130-5G
76895-280EA 168.69 USD
76895-280 76895-282 76895-284 76895-286
2-Chloro-5-methylnicotinaldehyde ≥97%
2-Chloro-5-methylnicotinaldehyde
Formula: C₇H₆ClNO
MW: 155.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375370
CAS Number: 92444-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR