3-Formyl-2,6-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethoxynicotinaldehyde, 2,6-Dimethoxypyridine-3-carboxaldehyde

14123-25G 14123-5G
76895-270EA 315.09 USD
76895-270 76895-272
3-Formyl-2,6-dimethoxypyridine ≥97%
3-Formyl-2,6-dimethoxypyridine
Formula: C₈H₉NO₃
MW: 167.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064048
CAS Number: 58819-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR