2-Formyl-5-(trifluoromethoxy)benzoic acid ≥97%
Supplier: AOBChem USA
14111-1G
14111-25G
14111-500MG
14111-5G
76895-254EA
441.34
USD
76895-254
76895-256
76895-258
76895-260
2-Formyl-5-(trifluoromethoxy)benzoic acid ≥97%
2-Formyl-5-(trifluoromethoxy)benzoic acid
Formula:
C₉H₅F₃O₄ |
MDL Number:
MFCD28976756 CAS Number: 2167892-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...