2-Bromo-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14043-1G 14043-25G 14043-5G
76895-214EA 96.55 USD
76895-214 76895-216 76895-218
2-Bromo-4-(trifluoromethoxy)benzaldehyde ≥97%
2-Bromo-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09835093
CAS Number: 1114808-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR