(2-Hydroxy-3-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

14009-1G 14009-25G 14009-5G
76895-204EA 64.72 USD
76895-204 76895-206 76895-208
(2-Hydroxy-3-methylphenyl)boronic acid ≥97%
(2-Hydroxy-3-methylphenyl)boronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689549
CAS Number: 259209-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR