2-Methyl-1-(3-(trifluoromethoxy)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139879-1G 139879-25G 139879-500MG 139879-5G
76895-192EA 263.11 USD
76895-192 76895-194 76895-196 76895-198
2-Methyl-1-(3-(trifluoromethoxy)phenyl)propan-1-ol ≥95%
2-Methyl-1-(3-(trifluoromethoxy)phenyl)propan-1-ol
Formula: C₁₁H₁₃F₃O₂
MW: 234.215 g/mol
Storage Temperature: Ambient
CAS Number: 1343182-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR