2-nitro-1-phenoxy-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

138941-1G 138941-25G 138941-500MG 138941-5G
76895-170EA 222.79 USD
76895-170 76895-172 76895-174 76895-176
2-nitro-1-phenoxy-4-(trifluoromethyl)benzene ≥97%
2-nitro-1-phenoxy-4-(trifluoromethyl)benzene
Formula: C₁₃H₈F₃NO₃
MW: 283.203 g/mol
Storage Temperature: Ambient
CAS Number: 1960-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR