2-Amino-5-fluorobenzeneboronic acidpinacol ester ≥97%

Supplier: AOBChem USA

Warning

13820-1G 13820-25G 13820-500MG 13820-5G
76895-142EA 204.76 USD
76895-142 76895-144 76895-146 76895-148
2-Amino-5-fluorobenzeneboronic acidpinacol ester ≥97%
2-Amino-5-fluorobenzeneboronic acidpinacol ester
Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11617916
CAS Number: 863578-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR