2,6-Difluoro-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoropyridine-3-boronic acid, pinacol ester, 2,6-Difluoropyridine-3-boronic acid Pinacol ester, 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2,6-Difluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

13564-25G 13564-5G
76895-092EA 240.83 USD
76895-092 76895-094
2,6-Difluoro-3-pyridineboronic acid pinacol ester ≥97%
2,6-Difluoro-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BF₂NO₂
MW: 241.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037475
CAS Number: 1072945-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR