2-Chloro-3-cyano-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

13559-1G 13559-25G 13559-500MG 13559-5G
76895-076EA 248.26 USD
76895-076 76895-078 76895-080 76895-082
2-Chloro-3-cyano-4-pyridineboronic acid pinacol ester ≥97%
2-Chloro-3-cyano-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09037479
CAS Number: 878194-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR