3-(2-Bromophenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13487-25G 13487-5G
76895-062EA 325.7 USD
76895-062 76895-064
3-(2-Bromophenylcarbamoyl)phenylboronic acid ≥97%
3-(2-Bromophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027206
CAS Number: 874288-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR