2-Phenoxyphenol ≥97%

Supplier: AOBChem USA

Warning

134658-1G 134658-25G 134658-500MG 134658-5G
76895-048EA 209 USD
76895-048 76895-050 76895-052 76895-054
2-Phenoxyphenol ≥97%
2-Phenoxyphenol
Formula: C₁₂H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00093034
CAS Number: 2417-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR