2-Chloro-6-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13404-1G 13404-25G 13404-5G
76895-024EA 88.06 USD
76895-024 76895-026 76895-028
2-Chloro-6-(trifluoromethyl)phenylboronic acid ≥97%
2-Chloro-6-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.3726 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09037968
CAS Number: 851756-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR