3,5-Dimethoxysalicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-hydroxy-3,5-dimethoxybenzaldehyde

133065-1G 133065-500MG 133065-5G
76895-016EA 542.12 USD
76895-016 76895-018 76895-020
3,5-Dimethoxysalicylaldehyde ≥97%
3,5-Dimethoxysalicylaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
Storage Temperature: Refrigerator
MDL Number: MFCD06740708
CAS Number: 65162-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR