2-Bromo-3-fluoro-1-(methoxymethoxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

131134-1G 131134-500MG 131134-5G
76894-992EA 891.16 USD
76894-992 76894-994 76894-996
2-Bromo-3-fluoro-1-(methoxymethoxy)-4-methylbenzene ≥97%
2-Bromo-3-fluoro-1-(methoxymethoxy)-4-methylbenzene
Formula: C₉H₁₀BrFO₂
MW: 249.0769 g/mol
Storage Temperature: Refrigerator
CAS Number: 2158298-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR