(2-Isopropoxypyrimidin-5-yl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Isopropoxy-5-pyrimidineboronic acid

12985-1G 12985-25G 12985-500MG 12985-5G
76894-976EA 130.5 USD
76894-976 76894-978 76894-980 76894-982
(2-Isopropoxypyrimidin-5-yl)boronic acid ≥97% research grade
(2-Isopropoxypyrimidin-5-yl)boronic acid
Formula: C₇H₁₁BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD16618883
CAS Number: 870521-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR