2-Bromo-6-chloro-4-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

128642-1G 128642-25G 128642-500MG 128642-5G
76894-926EA 207.94 USD
76894-926 76894-928 76894-930 76894-932
2-Bromo-6-chloro-4-methylpyridin-3-ol ≥97%
2-Bromo-6-chloro-4-methylpyridin-3-ol
Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Ambient
CAS Number: 1823370-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR