2-Hydroxymethyl-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12771-1G 12771-25G 12771-5G
76894-914EA 64.72 USD
76894-914 76894-916 76894-918
2-Hydroxymethyl-4-methoxyphenylboronic acid ≥97%
2-Hydroxymethyl-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MW: 181.982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566595
CAS Number: 762263-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR