2-Bromo-3,4-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

126823-1G 126823-25G 126823-500MG 126823-5G
76894-882EA 343.74 USD
76894-882 76894-884 76894-886 76894-888
2-Bromo-3,4-difluorophenol ≥97%
2-Bromo-3,4-difluorophenol
Formula: C₆H₃BrF₂O
Storage Temperature: Refrigerator
CAS Number: 1376335-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR