2-Phenylethylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
12525-25G
12525-5G
76894-874EA
268.41
USD
76894-874
76894-876
2-Phenylethylboronic acid pinacol ester ≥97%
2-Phenylethylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₂₁BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03788721 CAS Number: 165904-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...