2-(Methylthio)-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(Methylthio)pyridine-3-boronic acid, 2-Methylthiopyridine-5-boronic acid, 6-(Methylthio)pyridin-3-ylboronic acid

12482-1G 12482-25G 12482-5G
76894-862EA 84.88 USD
76894-862 76894-864 76894-866
2-(Methylthio)-5-pyridineboronic acid ≥97%
2-(Methylthio)-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD03788238
CAS Number: 321438-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR