2-Chloro-4-ethylphenol ≥97%

Supplier: AOBChem USA

Warning

123735-1G 123735-25G 123735-500MG 123735-5G
76894-846EA 213.25 USD
76894-846 76894-848 76894-850 76894-852
2-Chloro-4-ethylphenol ≥97%
2-Chloro-4-ethylphenol
Formula: C₈H₉ClO
Storage Temperature: Ambient
CAS Number: 18980-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR