2-Methoxy-4-(trifluoromethoxy)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-4-(trifluoromethoxy)phenylboronic acid, (2-Methoxy-4-(trifluoromethoxy)phenyl)boronic acid

12218-1G 12218-25G 12218-5G
76894-828EA 106.09 USD
76894-828 76894-830 76894-832
2-Methoxy-4-(trifluoromethoxy)benzeneboronic acid ≥97%
2-Methoxy-4-(trifluoromethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₄
MW: 235.953 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037221
CAS Number: 355836-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR