2-Methoxy-4-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Methoxy-4-(trifluoromethyl)phenyl)boronic acid

12207-25G 12207-5G
76894-820EA 418 USD
76894-820 76894-822
2-Methoxy-4-(trifluoromethyl)phenylboronic acid ≥95%
2-Methoxy-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.9500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03428511
CAS Number: 312936-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR