(2-(Methoxymethyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-methoxymethylphenyl)boronic acid, 2-(Methoxymethyl)phenylboronic acid, 2-(Methoxymethyl)benzeneboronic acid, 2-Methoxymethylphenylboronic acid

12164-25G 12164-5G
76894-814EA 212.18 USD
76894-814 76894-816
(2-(Methoxymethyl)phenyl)boronic acid ≥95%
(2-(Methoxymethyl)phenyl)boronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412110
CAS Number: 126617-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR