2-Isopropoxy-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12133-25G 12133-5G
76894-798EA 275.84 USD
76894-798 76894-800
2-Isopropoxy-5-methylphenylboronic acid ≥97%
2-Isopropoxy-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974067
CAS Number: 480438-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR