2-Isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Isopropoxybenzeneboronic acid, (2-Isopropoxyphenyl)boronic acid

12122-25G 12122-5G
76894-788EA 177.18 USD
76894-788 76894-790
2-Isopropoxyphenylboronic acid ≥97%
2-Isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD03427050
CAS Number: 138008-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR