2-Bromo-1-Ethoxy-4-Fluoro-3-(Methylsulfonyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

120248-1G 120248-500MG 120248-5G 120248-25G
76894-748EA 527.27 USD
76894-748 76894-750 76894-752 77657-278
2-Bromo-1-Ethoxy-4-Fluoro-3-(Methylsulfonyl)Benzene ≥97%
2-Bromo-1-Ethoxy-4-Fluoro-3-(Methylsulfonyl)Benzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₃S
Storage Temperature: Ambient
MDL Number: MFCD34784650
CAS Number: 2918906-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR