2-Formyl-5-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
119958-1G
119958-25G
119958-500MG
119958-5G
76894-722EA
390.42
USD
76894-722
76894-724
76894-726
76894-728
2-Formyl-5-methylbenzoic acid ≥97%
2-Formyl-5-methylbenzoic acid
Formula:
C₉H₈O₃ MW: 164.158 g/mol Storage Temperature: Refrigerator |
CAS Number:
150867-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...