2-Ethoxy-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11927-25G 11927-5G
76894-700EA 212.18 USD
76894-700 76894-702
2-Ethoxy-4-fluorophenylboronic acid ≥97%
2-Ethoxy-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD03095001
CAS Number: 480438-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR